Assay Details
| Gene Symbol: C29H11orf68 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105480.2 | NP_001098950.2 | 1 - 2 | 152 | 111 | |
| GenBank mRNA | BC134485.1 | - | 1 - 2 | 126 | 111 | |
| EV685608.1 | - | 1 - 2 | 155 | 111 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC134485.1 transcripts.
Set Membership: |
|

C29H11orf68
Genomic Map