Assay Details
| Gene Symbol: KAT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105483.2 | NP_001098953.1 | 1 - 2 | 224 | 53 | |
| GenBank mRNA | BC133611.1 | - | 1 - 2 | 226 | 53 | |
More Information
Set Membership: |
|

KAT8
Genomic Map