Assay Details
| Gene Symbol: SCO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105493.2 | NP_001098963.1 | 1 - 2 | 481 | 107 | |
| GenBank mRNA | BC146150.1 | - | 1 - 2 | 482 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC146150.1, NM_001105493.2 transcripts.
Set Membership: |
|

SCO2
Genomic Map