Assay Details
| Gene Symbol: ELOF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105508.2 | NP_001098978.1 | 1 - 2 | 50 | 74 | |
| GenBank mRNA | BC134722.1 | - | 1 - 2 | 51 | 74 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC134722.1, NM_001105508.2 transcripts.
Set Membership: |
|

ELOF1
Genomic Map