Assay Details
| Gene Symbol: ELOF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105508.2 | NP_001098978.1 | 3 - 4 | 253 | 61 | |
| GenBank mRNA | BC134722.1 | - | 3 - 4 | 254 | 61 | |
More Information
Set Membership: |
|

ELOF1
Genomic Map