Assay Details
| Gene Symbol: COX19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109966.1 | NP_001103436.1 | 2 - 3 | 213 | 78 | |
| GenBank mRNA | BC134520.1 | - | 2 - 3 | 213 | 78 | |
More Information
Set Membership: |
|

COX19
Genomic Map