Assay Details
| Gene Symbol: ZHX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110095.1 | NP_001103565.1 | 3 - 4 | 3026 | 96 | |
| GenBank mRNA | BC154375.1 | - | 3 - 4 | 3026 | 96 | |
More Information
Set Membership: |
|

ZHX1
Genomic Map