Assay Details
| Gene Symbol: CD302 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110191.1 | NP_001103661.1 | 1 - 2 | 88 | 133 | |
| GenBank mRNA | BC105308.1 | - | 1 - 2 | 88 | 133 | |
More Information
Set Membership: |
|

CD302
Genomic Map