Assay Details
| Gene Symbol: ZNF713 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110444.2 | NP_001103914.1 | 5 - 6 | 930 | 121 | |
| GenBank mRNA | BC126697.1 | - | 5 - 6 | 931 | 121 | |
More Information
Set Membership: |
|

ZNF713
Genomic Map