Assay Details
| Gene Symbol: LAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174098.3 | NP_776523.2 | 8 - 9 | 1062 | 61 | |
| GenBank mRNA | BC105385.1 | - | 8 - 9 | 965 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LAP3| Gene Symbol: LAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_174098.3 | NP_776523.2 | 8 - 9 | 1062 | 61 | |
| GenBank mRNA | BC105385.1 | - | 8 - 9 | 965 | 61 | |
Set Membership: |
|