Assay Details
| Gene Symbol: KRT83 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083385.1 | NP_001076854.1 | 4 - 5 | 797 | 136 | |
| GenBank mRNA | BC123471.1 | - | 4 - 5 | 797 | 136 | |
More Information
Set Membership: |
|

KRT83
Genomic Map