Assay Details
| Gene Symbol: BOLA-NC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105616.2 | NP_001099086.1 | 1 - 2 | 71 | 62 | |
| GenBank mRNA | AY188807.2 | - | 1 - 2 | 51 | 62 | |
| BC109706.1 | - | 1 - 2 | 72 | 62 | ||
| DQ140369.1 | - | 1 - 2 | 46 | 62 | ||
| DQ140370.1 | - | 1 - 2 | 46 | 62 | ||
| KT428358.1 | - | 1 - 2 | 51 | 62 | ||
More Information
Set Membership: |
|

BOLA-NC1
Genomic Map