Assay Details
| Gene Symbol: BOLA-NC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105616.2 | NP_001099086.1 | 2 - 3 | 346 | 115 | |
| GenBank mRNA | BC109706.1 | - | 2 - 3 | 347 | 115 | |
| DQ140369.1 | - | 2 - 3 | 321 | 115 | ||
| DQ140370.1 | - | 2 - 3 | 321 | 115 | ||
| KT428358.1 | - | 2 - 3 | 326 | 115 | ||
| KT428359.1 | - | 2 - 3 | 321 | 115 | ||
| KT428361.1 | - | 2 - 3 | 321 | 115 | ||
| KT428362.1 | - | 2 - 3 | 321 | 115 | ||
| KT428363.1 | - | 2 - 3 | 321 | 115 | ||
More Information
Set Membership: |
|

BOLA-NC1
Genomic Map