Assay Details
| Gene Symbol: KRT33A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099099.1 | NP_001092569.1 | 6 - 7 | 1148 | 144 | |
| GenBank mRNA | BC142131.1 | - | 6 - 7 | 1148 | 144 | |
More Information
Set Membership: |
|

KRT33A
Genomic Map