Assay Details
| Gene Symbol: H3-5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099370.2 | NP_001092840.1 | 1 - 2 | 477 | 131 | |
| GenBank mRNA | BC142372.1 | - | 1 - 2 | 478 | 131 | |
More Information
Set Membership: |
|

H3-5
Genomic Map