Assay Details
| Gene Symbol: CSTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100362.2 | NP_001093832.1 | 1 - 2 | 146 | 133 | |
| GenBank mRNA | BC149561.1 | - | 1 - 2 | 147 | 133 | |
More Information
Set Membership: |
|

CSTB
Genomic Map