Assay Details
| Gene Symbol: PRG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098472.1 | NP_001091942.1 | 5 - 6 | 712 | 139 | |
| GenBank mRNA | BC134800.1 | - | 5 - 6 | 712 | 139 | |
More Information
Set Membership: |
|

PRG3
Genomic Map