Assay Details
| Gene Symbol: PLA2G2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025324.2 | NP_001020495.1 | 2 - 3 | 113 | 165 | |
| GenBank mRNA | AY911335.1 | - | 2 - 3 | 114 | 165 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001025324.2 transcripts.
Set Membership: |
|

PLA2G2A
Genomic Map