Assay Details
| Gene Symbol: PLA2G2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001025324.2 | NP_001020495.1 | 3 - 4 | 262 | 125 | |
| GenBank mRNA | AY911335.1 | - | 3 - 4 | 263 | 125 | |
More Information
Set Membership: |
|

PLA2G2A
Genomic Map