Assay Details
| Gene Symbol: OAS1Z | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029846.2 | NP_001025017.2 | 3 - 4 | 784 | 149 | |
| GenBank mRNA | AY650038.1 | - | 3 - 4 | 716 | 149 | |
More Information
Set Membership: |
|

OAS1Z
Genomic Map