Assay Details
| Gene Symbol: GSTA2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_177515.2 | NP_803481.1 | 6 - 7 | 579 | 158 | |
| GenBank mRNA | AF027386.1 | - | 6 - 7 | 579 | 158 | |
| BC114835.1 | - | 6 - 7 | 645 | 158 | ||
More Information
Set Membership: |
|

GSTA2
Genomic Map