Assay Details
| Gene Symbol: LYZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077829.1 | NP_001071297.1 | 1 - 2 | 160 | 126 | |
| GenBank mRNA | AY684064.1 | - | 1 - 2 | 160 | 126 | |
| BC103297.1 | - | 1 - 2 | 151 | 126 | ||
More Information
Set Membership: |
|

LYZ1
Genomic Map