Assay Details
| Gene Symbol: UTP14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034241.2 | NP_001029413.1 | 1 - 2 | 70 | 126 | |
| GenBank mRNA | BC102296.1 | - | 1 - 2 | 71 | 126 | |
| BT030541.1 | - | 1 - 2 | 57 | 126 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102296.1, NM_001034241.2 transcripts.
Set Membership: |
|

UTP14A
Genomic Map