Assay Details
| Gene Symbol: UTP14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034241.2 | NP_001029413.1 | 2 - 3 | 150 | 144 | |
| GenBank mRNA | BC102296.1 | - | 2 - 3 | 151 | 144 | |
| BT030541.1 | - | 2 - 3 | 137 | 144 | ||
More Information
Set Membership: |
|

UTP14A
Genomic Map