Assay Details
| Gene Symbol: UTP14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034241.2 | NP_001029413.1 | 7 - 8 | 701 | 136 | |
| GenBank mRNA | BC102296.1 | - | 7 - 8 | 702 | 136 | |
| BT030541.1 | - | 7 - 8 | 688 | 136 | ||
More Information
Set Membership: |
|

UTP14A
Genomic Map