Assay Details
| Gene Symbol: TUBG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037615.2 | NP_001032704.1 | 1 - 2 | 195 | 97 | |
| GenBank mRNA | BC110028.1 | - | 1 - 2 | 196 | 97 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC110028.1, NM_001037615.2 transcripts.
Set Membership: |
|

TUBG2
Genomic Map