Assay Details
| Gene Symbol: TUBG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037615.2 | NP_001032704.1 | 4 - 5 | 539 | 123 | |
| GenBank mRNA | BC110028.1 | - | 4 - 5 | 540 | 123 | |
More Information
Set Membership: |
|

TUBG2
Genomic Map