Assay Details
| Gene Symbol: KRT31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076506.1 | NP_001069974.1 | 2 - 3 | 478 | 149 | |
| GenBank mRNA | BC118288.1 | - | 2 - 3 | 478 | 149 | |
More Information
Set Membership: |
|

KRT31
Genomic Map