Assay Details
Gene Symbol: DEFB122 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001078107.2 | NP_001071575.1 | 1 - 2 | 111 | 147 | |
GenBank mRNA | BC120465.1 | - | 1 - 2 | 112 | 147 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC120465.1, NM_001078107.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
