Assay Details
| Gene Symbol: LYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078159.1 | NP_001071627.1 | 1 - 2 | 163 | 121 | |
| GenBank mRNA | BC112597.1 | - | 1 - 2 | 163 | 121 | |
| HQ285242.1 | - | 1 - 2 | 133 | 121 | ||
| L19980.1 | - | 1 - 2 | 114 | 121 | ||
| L23757.1 | - | 1 - 2 | 143 | 121 | ||
More Information
Set Membership: |
|

LYZ
Genomic Map