Assay Details
| Gene Symbol: LYZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078159.1 | NP_001071627.1 | 3 - 4 | 411 | 156 | |
| GenBank mRNA | BC112597.1 | - | 3 - 4 | 411 | 156 | |
| HQ285242.1 | - | 3 - 4 | 381 | 156 | ||
| L19980.1 | - | 3 - 4 | 362 | 156 | ||
| L23757.1 | - | 3 - 4 | 391 | 156 | ||
More Information
Set Membership: |
|

LYZ
Genomic Map