Assay Details
| Gene Symbol: KCNB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024563.1 | NP_001019734.1 | 3 | 2750 | 114 | |
| GenBank mRNA | DQ009866.1 | - | 3 | 2750 | 114 | |
More Information
Set Membership: |
|

KCNB2
Genomic Map