Assay Details
| Gene Symbol: TMEM164 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001031762.1 | NP_001026932.1 | 4 | 747 | 147 | |
| GenBank mRNA | BC120259.1 | - | 7 | 919 | 147 | |
| BT020678.1 | - | 4 | 747 | 147 | ||
More Information
Set Membership: |
|

TMEM164
Genomic Map