Assay Details
| Gene Symbol: LFNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046222.2 | NP_001039687.1 | 8 | 1855 | 77 | |
| GenBank mRNA | BC105463.1 | - | 8 | 1856 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LFNG| Gene Symbol: LFNG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046222.2 | NP_001039687.1 | 8 | 1855 | 77 | |
| GenBank mRNA | BC105463.1 | - | 8 | 1856 | 77 | |
Set Membership: |
|