Assay Details
| Gene Symbol: LOC534967 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046391.1 | NP_001039856.1 | 13 | 1648 | 143 | |
| GenBank mRNA | BC113218.1 | - | 13 | 1648 | 143 | |
More Information
Set Membership: |
|

LOC534967
Genomic Map