Assay Details
| Gene Symbol: NFIL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075240.1 | NP_001068708.1 | 1 | 1646 | 147 | |
| GenBank mRNA | BC123885.1 | - | 1 | 1646 | 147 | |
More Information
Set Membership: |
|

NFIL3
Genomic Map