Assay Details
| Gene Symbol: CH25H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075243.1 | NP_001068711.1 | 1 | 1750 | 152 | |
| GenBank mRNA | BC120312.1 | - | 1 | 1750 | 152 | |
More Information
Set Membership: |
|

CH25H
Genomic Map