Assay Details
| Gene Symbol: MARCKS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076276.1 | NP_001069744.1 | 2 | 548 | 127 | |
| GenBank mRNA | BC115989.1 | - | 2 | 909 | 127 | |
More Information
Set Membership: |
|

MARCKS
Genomic Map