Assay Details
| Gene Symbol: LYRM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076306.1 | NP_001069774.1 | 3 | 845 | 129 | |
| GenBank mRNA | BC120188.1 | - | 3 | 845 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LYRM4| Gene Symbol: LYRM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076306.1 | NP_001069774.1 | 3 | 845 | 129 | |
| GenBank mRNA | BC120188.1 | - | 3 | 845 | 129 | |
Set Membership: |
|