Assay Details
| Gene Symbol: UCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076530.1 | NP_001069998.1 | 1 | 151 | 151 | |
| GenBank mRNA | AB252572.1 | - | 1 | 151 | 151 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UCN2| Gene Symbol: UCN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076530.1 | NP_001069998.1 | 1 | 151 | 151 | |
| GenBank mRNA | AB252572.1 | - | 1 | 151 | 151 | |
Set Membership: |
|