Assay Details
| Gene Symbol: FSCB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076546.2 | NP_001070014.1 | 1 | 2553 | 125 | |
| GenBank mRNA | BC111347.1 | - | 1 | 2554 | 125 | |
More Information
Set Membership: |
|

FSCB
Genomic Map