Assay Details
| Gene Symbol: CLEC14A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001077890.1 | NP_001071358.1 | 1 | 1743 | 90 | |
| GenBank mRNA | BC118138.1 | - | 1 | 1743 | 90 | |
More Information
Set Membership: |
|

CLEC14A
Genomic Map