Assay Details
| Gene Symbol: SOX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078128.1 | NP_001071596.1 | 1 | 754 | 114 | |
| GenBank mRNA | BC120191.1 | - | 1 | 754 | 114 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SOX4| Gene Symbol: SOX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001078128.1 | NP_001071596.1 | 1 | 754 | 114 | |
| GenBank mRNA | BC120191.1 | - | 1 | 754 | 114 | |
Set Membership: |
|