Assay Details
| Gene Symbol: PPP1R2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079599.2 | NP_001073067.1 | 1 | 752 | 108 | |
| GenBank mRNA | BC109613.1 | - | 1 | 753 | 108 | |
More Information
Set Membership: |
|

PPP1R2C
Genomic Map