Assay Details
| Gene Symbol: POU3F2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080348.1 | NP_001073817.1 | 1 | 1450 | 150 | |
| GenBank mRNA | BC112696.1 | - | 1 | 1450 | 150 | |
More Information
Set Membership: |
|

POU3F2
Genomic Map