Assay Details
| Gene Symbol: ZNF775 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080373.1 | NP_001073842.1 | 4 | 1351 | 97 | |
| GenBank mRNA | BC126698.1 | - | 4 | 1351 | 97 | |
More Information
Set Membership: |
|

ZNF775
Genomic Map