Assay Details
Gene Symbol: KCNJ10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001081601.1 | NP_001075070.1 | 2 | 4950 | 139 | |
GenBank mRNA | BC133349.1 | - | 2 | 139 | ||
EE226512.1 | - | 1 | 214 | 139 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
