Assay Details
| Gene Symbol: TSSK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083710.2 | NP_001077179.1 | 1 | 746 | 78 | |
| GenBank mRNA | BC102681.1 | - | 1 | 747 | 78 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TSSK1B| Gene Symbol: TSSK1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083710.2 | NP_001077179.1 | 1 | 746 | 78 | |
| GenBank mRNA | BC102681.1 | - | 1 | 747 | 78 | |
Set Membership: |
|