Assay Details
| Gene Symbol: PRP14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097562.1 | NP_001091031.1 | 1 | 51 | 83 | |
| GenBank mRNA | AB255602.1 | - | 1 | 51 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001097562.1 transcripts.
Set Membership: |
|

PRP14
Genomic Map