Assay Details
| Gene Symbol: TMPPE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099711.1 | NP_001093181.1 | 1 | 1547 | 146 | |
| GenBank mRNA | BC142143.1 | - | 1 | 1547 | 146 | |
More Information
Set Membership: |
|

TMPPE
Genomic Map